Learn More
Sino Biological Influenza A H3N2 (A/Hong Kong/4801/2014) Nucleoprotein / NP Protein (His Tag) 100ug
Supplier:  Sino Biological 40781V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Hong Kong/4801/2014 (H3N2)) nucleoprotein (translated amino acids of EPI653194) (Met1-Asn498) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4270