Learn More
Sino Biological Influenza A H3N2 (A/Minnesota/41/2019) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40848V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A H3N2 (A/Minnesota/41/2019) Hemagglutinin (EPI1487157) (Met1-Asp529) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6505