Learn More
Sino Biological Influenza A H3N2 (A/Perth/16/2009) Hemagglutinin / HA1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 40043T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H3N2 (A/Perth/16/2009) Hemagglutinin / HA1 ( Catalog#40043-V08H1; ACS71642.1; Met1-Arg345). Influenza A H3N2 (A/Perth/16/2009) Hemagglutinin / HA1 specific IgG was purified by Influenza A H3N2 (A/Perth/16/2009) Hemagglutinin / HA1 affinity chromatography.
Catalog No. 50-224-7335