Learn More
Sino Biological Influenza A H3N2 (A/Singapore/INFIMH-16-0019/2016) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40580V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A H3N2 (A/Singapore/INFIMH-16-0019/2016) Hemagglutinin (EPI1341068) (Met1-Trp530) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5711