Learn More
Sino Biological Influenza A H3N2 (A/swine/England/90591/1997) Hemagglutinin / HA Protein (His Tag) 100ug
Supplier:  Sino Biological 40685V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/swine/England/90591/1997 (H3N2)) hemagglutinin (AFR76481) (Met1-Ser525), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5807