Learn More
Sino Biological Influenza A H3N2 Neuraminidase / NA (Active)
Supplier:  Sino Biological 40017VNAHC800
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Babol/36/2005 (H3N2)) neuraminidase (ACN50232.1) (His 36-Pro 459) was expressed, the cell lysates are collected, and bio-activity was tested.
Catalog No. NC1917386