Learn More
Sino Biological Influenza A H7N9 (A/Anhui/1/2013) Neuraminidase / NA (His Tag), Biotinylated 20ug
Supplier:  Sino Biological 40108V07HB20
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Anhui/1/2013(H7N9)) neuraminidase (EPI439509) (His36-Leu465) was expressed with an N-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5608
 Restricted Item