Learn More
Sino Biological Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA Antibody (Biotin), Rabbit Pab 100ug
Supplier:  Sino Biological 40103RP01B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza A H7N9 (A/Shanghai/1/2013) Hemagglutinin / HA (Catalog#40103-V08B; EPI439507; Met1-Val524). Total IgG was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-224-6463