Learn More
Sino Biological Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA ELISA Pair Set 5 Plates
Supplier:  Sino Biological SEK401035
Encompass_Preferred
The Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA ELISA Pair Set is for the quantitative determination of Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA coated on a 96-well plate. Standards and samples are added to the wells, and any Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Influenza A H7N9 (A/Anhui/1/2013) Hemagglutinin / HA present in the sample. To end the enzyme reaction, the stop solution
Catalog No. 50-222-1506