Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Influenza A hemagglutinin (HA) protein, H6N2 (AFJ19048, sf9, His) | >95.0% | 50 UG
SDP

Catalog No. 5000290212
Encompass_Preferred

Recombinant Influenza A hemagglutinin (HA) protein, subtype H6N2, expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a purified protein for research use in immunoassays, antibody production, structural studies, and vaccine research. Certificate of analysis indicates purity >95% (reducing SDS-PAGE) and low endotoxin level (<1 EU/μg).

  • Recombinant protein expressed in Sf9 insect cells with C-terminal His tag.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Suitable for immunoassays, antibody generation, and vaccine research.
  • Provided with datasheet and certificate of analysis for lot-specific data.

Catalog No. 50-002-90212 Supplier Medchemexpress LLC Supplier No. HYP7645050UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Influenza A hemagglutinin (HA) protein, subtype H6N2, expressed in Sf9 insect cells with a C-terminal His tag. Supplied as a purified protein for research use in immunoassays, antibody production, structural studies, and vaccine research. Certificate of analysis indicates purity >95% (reducing SDS-PAGE) and low endotoxin level (<1 EU/μg).

  • Recombinant protein expressed in Sf9 insect cells with C-terminal His tag.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Suitable for immunoassays, antibody generation, and vaccine research.
  • Provided with datasheet and certificate of analysis for lot-specific data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.