Learn More
Creative Biomart Recombinant Influenza A H9N2 (Hong-Kong) Antigen
Catalog No. 502084855
Encompass_Preferred
Recombinant Full-Length H9N2 A/Hong-Kong/1073/99 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is 72,000 dalton. The accession number is AJ404626.
Catalog No. 50-208-4855 Supplier Creative Biomart Supplier No. VAC0015
 Restricted Item
Add to Cart
Add to Cart
Recombinant Full-Length H9N2 A/Hong-Kong/1073/99 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is 72,000 dalton. The accession number is AJ404626.