Learn More
Creative Biomart Recombinant Influenza A H5N1 (Indonesia) Antigen
Supplier:  Creative Biomart VAC0013
Encompass_Preferred
Recombinant Full-Length H5N1 A/Indonesia/05/2005 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells.
Catalog No. 50-208-4853