Learn More
Creative Biomart Recombinant Influenza A H3N2 (Wyoming) Antigen
Supplier:  Creative Biomart VAC0012
Encompass_Preferred
Recombinant Full-Length H3N2 A/Wyoming/2003/3 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is 72,000 Dalton.Accession number: AY531033.
Catalog No. 50-208-4852