Learn More
Creative Biomart Thioredoxin
Supplier:  Creative Biomart VAC0355
Encompass_Preferred
Thioredoxin is a small dithiol protein and specific reductant for major allergenic proteins present in foods from animal and plant products. It is a 12-kD oxidoreductase enzyme encoded by TXN and TXN2
Catalog No. 50-208-5195