Learn More
Creative Biomart Recombinant Mycoplasma Pneumoniae P1 Antigen, His-tagged
Supplier:  Creative Biomart VAC0228
Encompass_Preferred
The recombinant Mycoplasma Pneumoniae C-terminal region (P1C) of the P1 protein was expressed in E. coli containing 362 amino acids. The protein is fused to a 6 His Tag.This peptide can be used as
Catalog No. 50-208-5068