Learn More
Creative Biomart Recombinant HIV-1, 2 Antigen
Supplier:  Creative Biomart VAC0222
Encompass_Preferred
HIV-1,2 recombinant-E. coli derived recombinant 27 kDa protein contains the C-terminus of gp120 and most of gp41. The protein is conjugated to a 23 amino acids synthetic peptide derived from gp39 of H
Catalog No. 50-208-5062
 Restricted Item