Learn More
Creative Biomart Recombinant HIV-2 Protease Antigen
Catalog No. 502085063
Encompass_Preferred
HIV-2 protease is an active homodimer having a molecular mass of 10.7kDa.
Catalog No. 50-208-5063 Supplier Creative Biomart Supplier No. VAC0223
 Restricted Item
Add to Cart
Add to Cart
HIV-2 protease is an active homodimer having a molecular mass of 10.7kDa.