Learn More
Creative Biomart Recombinant HIV-1 Integrase Antigen, His-tagged
Catalog No. 502085056
Encompass_Preferred
The E. coli derived 36 kDa recombinant protein is a non-glycosylated polypeptide chain, containing the HIV-1 immunodominant regions from the pol protein (intergrase) and fused with a six histidines ta
Catalog No. 50-208-5056 Supplier Creative Biomart Supplier No. VAC0216
The E. coli derived 36 kDa recombinant protein is a non-glycosylated polypeptide chain, containing the HIV-1 immunodominant regions from the pol protein (intergrase) and fused with a six histidines ta