Learn More
Creative Biomart Recombinant HIV-1 nef Antigen, beta-galactosidase-tagged
Supplier:  Creative Biomart VAC0217
Encompass_Preferred
The E. coli derived 20 kDa recombinant protein genotype IIIB is a non-glycosylated polypeptide chain, containing the HIV-1 nef immunodominant regions, 3-190 amino acids. The HIV-1 nef is fused to beta
Catalog No. 50-208-5057