Learn More
Creative Biomart Recombinant HIV-1 gp41 Antigen, B-galactosidase-tagged
Supplier:  Creative Biomart VAC0214
Encompass_Preferred
HIV-1 env gp41 Strain IIIB is a non-glycosylated 288 amino acids polypeptide chain (a.a. 466-753) having a molecular mass of 32kDa. The protein is fused to B-galactosidase (114 kDa) at the N-terminus.
Catalog No. 50-208-5054