Learn More
Creative Biomart Recombinant HIV-1 gp120 CM Antigen
Supplier:  Creative Biomart VAC0212
Encompass_Preferred
HIV-1 gp120 CM Recombinant-is the external envelope protein, full-length 120 kDa, derived from the env. gene of HIV-1 and glycosylated with N-linked sugars and produced using baculovirus vectors in in
Catalog No. 50-208-5052