Learn More
ATCC Influenza A virus (H1N1); A/California/04/2009
Catalog No. 502382349
Encompass
Influenza A virus (H1N1)pdm09 strain A/California/04/2009 (H1N1)pdm09 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated from a 10-year-old boy with asthma in San Diego with asthma in 2009 and was deposited by the California Department of Health. It can be used in respiratory disease research.
Catalog No. 50-238-2349 Supplier ATCC Supplier No. VR1805
Influenza A virus (H1N1)pdm09 strain A/California/04/2009 (H1N1)pdm09 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated from a 10-year-old boy with asthma in San Diego with asthma in 2009 and was deposited by the California Department of Health. It can be used in respiratory disease research.