Learn More
ATCC Influenza A virus (H1N1); A/FM/1/47
Catalog No. 502382130
Encompass
Influenza A virus (H1N1) strain A/FM/1/47 is propagated in SPF chicken embryonated eggs. This strain was isolated in 1947 from a human patient at Fort Monmout in New Jersey. This product has applications in influenza research.
Catalog No. 50-238-2130 Supplier ATCC Supplier No. VR97
Influenza A virus (H1N1) strain A/FM/1/47 is propagated in SPF chicken embryonated eggs. This strain was isolated in 1947 from a human patient at Fort Monmout in New Jersey. This product has applications in influenza research.