Learn More
ATCC Influenza A virus (H1N1); A/Mal/302/54
Catalog No. 502383292
Encompass
Influenza A virus (H1N1) strain A/Mal/302/54 is propagated in SPF chicken embryonated eggs with intra-allantoic inoculation. This strain was isolated in 1954 from a human patient in Malaya. This product has applications in emerging infectious disease, respiratory disease, and influenza research.
Catalog No. 50-238-3292 Supplier ATCC Supplier No. VR98
Influenza A virus (H1N1) strain A/Mal/302/54 is propagated in SPF chicken embryonated eggs with intra-allantoic inoculation. This strain was isolated in 1954 from a human patient in Malaya. This product has applications in emerging infectious disease, respiratory disease, and influenza research.