Learn More
ATCC Influenza A virus (H1N1); A/NWS/33
Catalog No. 502382962
Encompass
Influenza A virus (H1N1) strain A/NWS/33 is propagated in SPF chicken embryonated eggs. This is a neurotropic strain of Influenza A virus that was derived from strain A/WS/33 by intracerebral inoculation of mice. It has applications in influenza and respiratory disease research.
Catalog No. 50-238-2962 Supplier ATCC Supplier No. VR219
Influenza A virus (H1N1) strain A/NWS/33 is propagated in SPF chicken embryonated eggs. This is a neurotropic strain of Influenza A virus that was derived from strain A/WS/33 by intracerebral inoculation of mice. It has applications in influenza and respiratory disease research.