Learn More
ATCC Influenza A virus (H1N1) strain A/PR/8/34 (ATCC VR-1469™)
Catalog No. 502382886
Encompass
Influenza A virus (H1N1) strain A/PR/8/34 (ATCC VR-1469™) is a tissue culture-adapted strain propagated in MDCK cells. Originally isolated in 1934 from a human patient in Puerto Rico, it is used in respiratory disease research and assay development.
Recommended companion products:
MDCK cells (Fisher Cat. No. 50-238-3399, ATCC CCL-34™)
EMEM (Fisher Cat. No. 50-238-2632, ATCC 30-2003™).
Catalog No. 50-238-2886 Supplier ATCC Supplier No. VR1469
Influenza A virus (H1N1) strain A/PR/8/34 (ATCC VR-1469™) is a tissue culture-adapted strain propagated in MDCK cells. Originally isolated in 1934 from a human patient in Puerto Rico, it is used in respiratory disease research and assay development.
Recommended companion products:
MDCK cells (Fisher Cat. No. 50-238-3399, ATCC CCL-34™)
EMEM (Fisher Cat. No. 50-238-2632, ATCC 30-2003™).