Learn More
ATCC Influenza A virus (H1N1) strain A/PR/8/34 (ATCC VR-95™)
Catalog No. 502382155
Encompass
Influenza A virus (H1N1) strain A/PR/8/34 (ATCC VR-95™) is a whole-genome sequenced virus originally isolated in 1934 from a human patient in Puerto Rico. Propagated in SPF chicken embryonated eggs, it is widely used in respiratory disease and influenza research.
Recommended companion product:
Dulbecco's Phosphate Buffered Saline (D-PBS), 1X (Fisher Cat. No. 50-238-4473, ATCC 30-2200™).
Catalog No. 50-238-2155 Supplier ATCC Supplier No. VR95
Influenza A virus (H1N1) strain A/PR/8/34 (ATCC VR-95™) is a whole-genome sequenced virus originally isolated in 1934 from a human patient in Puerto Rico. Propagated in SPF chicken embryonated eggs, it is widely used in respiratory disease and influenza research.
Recommended companion product:
Dulbecco's Phosphate Buffered Saline (D-PBS), 1X (Fisher Cat. No. 50-238-4473, ATCC 30-2200™).