Learn More
ATCC Influenza A virus (H1N1); A/Virginia/ATCC3/2009; TC isolate
Catalog No. 502382261
Encompass
Influenza A virus (H1N1) strain A/Virginia/ATCC3/2009 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated in 2009 from a nasopharyngeal specimen from a patient positive for Influenza A in Virginia. It has applications in respiratory research and influenza research.
Catalog No. 50-238-2261 Supplier ATCC Supplier No. VR1738
Influenza A virus (H1N1) strain A/Virginia/ATCC3/2009 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated in 2009 from a nasopharyngeal specimen from a patient positive for Influenza A in Virginia. It has applications in respiratory research and influenza research.