Learn More
ATCC Influenza A virus (H1N1); A/Virginia/ATCC2/2009; RNA
Catalog No. 502385064
Encompass
Genomic RNA isolated from a preparation of cell lysate and supernatant from MDCK (ATCC CCL-34) cells infected with Influenza A virus (H1N1) strain A/Virginia/ATCC2/2009 (ATCC VR-1737). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-5064 Supplier ATCC Supplier No. VR1737D
Genomic RNA isolated from a preparation of cell lysate and supernatant from MDCK (ATCC CCL-34) cells infected with Influenza A virus (H1N1) strain A/Virginia/ATCC2/2009 (ATCC VR-1737). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.