Learn More
ATCC Influenza A virus (H1N1); A/WS/33; TC adapted
Catalog No. 502384316
Encompass
Influenza A virus (H1N1) strain A/WS/33 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated from the throat washings from an influenza patient and was deposited by the Centers for Disease Control and Prevention. It has applications in respiratory disease research.
Catalog No. 50-238-4316 Supplier ATCC Supplier No. VR1520
Influenza A virus (H1N1) strain A/WS/33 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated from the throat washings from an influenza patient and was deposited by the Centers for Disease Control and Prevention. It has applications in respiratory disease research.