Learn More
Bioworld INFLUENZA A VIRUS H1N1 BEIJ
Supplier:  Bioworld 220605151
Encompass_Preferred
INFLUENZA A VIRUS H1N1 BEIJING 262/95 (RECOMBINANT): Recombinant Hemagglutinin H1N1 New Caledonia 20/99 Source: Insect cells Description : Recombinant Hemagglutinin is glycosylated with N-linked sugars, produced using baculovirus vec...
Catalog No. 50-253-613