Learn More
ATCC Influenza A virus (H1N1pdm); A/California/07/2009NYMC X-179A
Catalog No. 502382327
Encompass
Influenza A virus (H1N1pdm) strain A/California/07/2009 NYMC X-179A is propagated in SPF embryonated chicken eggs. This strain is a classical reassortant virus derived from A/California/07/2009 (H1N1)pdm09 and A/Puerto Rico/8/1934 (H1N1). It has applications in infectious and respiratory disease research and in vaccine development.
Catalog No. 50-238-2327 Supplier ATCC Supplier No. VR1884
Influenza A virus (H1N1pdm) strain A/California/07/2009 NYMC X-179A is propagated in SPF embryonated chicken eggs. This strain is a classical reassortant virus derived from A/California/07/2009 (H1N1)pdm09 and A/Puerto Rico/8/1934 (H1N1). It has applications in infectious and respiratory disease research and in vaccine development.