Learn More
ATCC Influenza A virus (H3N2); A/Aichi/2/68; RNA
Catalog No. 502384392
Encompass
Genomic RNA isolated from MDCK cells (ATCC CCL-34) infected with Influenza A virus (H3N2) strain A/Aichi/2/68 (ATCC VR-1680). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.
Catalog No. 50-238-4392 Supplier ATCC Supplier No. VR1680D
Genomic RNA isolated from MDCK cells (ATCC CCL-34) infected with Influenza A virus (H3N2) strain A/Aichi/2/68 (ATCC VR-1680). This product was prepared using methods known to inactivate viruses. It is suitable for use in RT-PCR or other RNA-based procedures. The source organism and host cells are also available through the ATCC catalog.