Learn More
ATCC Influenza A virus (H3N2); A/Aichi/2/68; Cell culture adapted
Catalog No. 502383882
Encompass
Influenza A virus (H3N2) strain A/Aichi/2/68 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated in 1968 from a sailor on an Israeli ship docking in Aichi, Japan, and was deposited by the Centers for Disease Control and Prevention. It can be used in respiratory disease and influenza research.
Catalog No. 50-238-3882 Supplier ATCC Supplier No. VR1680
Influenza A virus (H3N2) strain A/Aichi/2/68 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated in 1968 from a sailor on an Israeli ship docking in Aichi, Japan, and was deposited by the Centers for Disease Control and Prevention. It can be used in respiratory disease and influenza research.