Learn More
ATCC Influenza A virus (H3N2); A/Victoria/3/75
Catalog No. 502384714
Encompass
Influenza A virus (H3N2) strain A/Victoria/3/75 is propagated in 10-day-old SPF chicken embryonated eggs. This strain was isolated from a human patient in Melbourne, Australia, and was deposited by AP Kendal. This product is whole-genome sequenced and has applications in influenza research.
Catalog No. 50-238-4714 Supplier ATCC Supplier No. VR822
Influenza A virus (H3N2) strain A/Victoria/3/75 is propagated in 10-day-old SPF chicken embryonated eggs. This strain was isolated from a human patient in Melbourne, Australia, and was deposited by AP Kendal. This product is whole-genome sequenced and has applications in influenza research.