Learn More
Bioworld Influenza A Virus H3N2 Panama 2007/99 (Recombinant), 10 μg
Supplier:  Bioworld 220605191
Encompass_Preferred
Influenza virus, type A (H3N2) Panama/2007/99 Source: Allantoic fluid Purification: Purified by ultracentrifugation using 10-40% sucrose gradient. Presentation: Solution in STE containing 0.1% NaN3 and 0.005% merthiolate. Inactivation: 0.005% merthiolate. This product has been treated in a manner consistent with methods of inactivation. Generally accepted good laboratory practices appropriate to microbiological/viral safe handling practices and techniques is required at work. Analysis: Purity more than 90% according to 7.5 % SDS-PAGE. Immunological activity: Has been tested with anti-influenza A monoclonal antibodies in ELISA Application: Serological studies of influenza A virus, immunogen for antibody production. Storage: - 20 C. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-617