Learn More
ATCC Influenza A virus (H3N2); RNA; A/Virginia/ATCC6/2012
Catalog No. 502384769
Encompass
Genomic RNA isolated from Influenza A virus (H3N2) strain A/Virginia/ATCC6/2012 (ATCC VR-1811). This product was prepared using methods known to inactivate viruses. It can be used in PCR and other molecular biology applications.
Catalog No. 50-238-4769 Supplier ATCC Supplier No. VR1811D
Genomic RNA isolated from Influenza A virus (H3N2) strain A/Virginia/ATCC6/2012 (ATCC VR-1811). This product was prepared using methods known to inactivate viruses. It can be used in PCR and other molecular biology applications.