Learn More
ATCC Influenza A virus(H3N2); A/Switzerland/9715293/2013
Supplier:  ATCC VR1837
Encompass
Influenza A virus (H3N2) strain A/Switzerland/9715293/2013 is propagated in SPF embryonated chicken eggs. This viral strain was isolated from a human patient in Switzerland in 2013. It has applications in influenza and respiratory disease research.
Catalog No. 50-238-4169