Learn More
Sino Biological Influenza B (B/Austria/1359417/2021) Neuraminidase / NA Protein (ECD, His Tag)
Supplier:  Sino Biological 40863V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza B (B/Austria/1359417/2021) Neuraminidase (EPI1868374) (Tyr33-Leu466) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and polyhistidine tag at the C-terminus.
Catalog No. 50-224-6518