Learn More
Sino Biological Influenza B (B/Austria/1359417/2021) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40862V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza B (B/Austria/1359417/2021) Hemagglutinin (EPI1868375) (Met1-Thr545) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6517