Learn More
Sino Biological Influenza B (B/Washington/02/2019) Nucleoprotein / NP Protein (His Tag)
Supplier:  Sino Biological 40755V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza B (B/Washington/02/2019) nucleoprotein (translated amino acids of EPI1368865) (Met1-Tyr560) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6002