Learn More
Sino Biological Influenza B (B/Washington/02/2019) Neuraminidase / NA (His Tag) 100ug
Supplier:  Sino Biological 40790V08B100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Washington/02/2019) neuraminidase (translated amino acids of EPI1394969) (Leu32-Leu466) was expressed with a vasodilator-stimulated phosphoprotein tetramerization domain at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-211-4237
 Restricted Item