Learn More
Sino Biological Influenza B Hemagglutinin / HA Antibody, Rabbit PAb
Supplier:  Sino Biological 11053RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Influenza B virus HA (Catalog#11053-V08H; ACA33493.1; Met 1-Ala 555). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1759