Learn More
Sino Biological Influenza B (B/Victoria/02/1987) Hemagglutinin / HA Protein (ECD, His Tag)
Supplier:  Sino Biological 40163V08B1100
Encompass_Preferred
A DNA sequence encoding the Influenza B (B/Victoria/02/1987) Hemagglutinin / HA Protein (ABL77244.1) (Met1-Thr548) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3739