Learn More
Sino Biological Influenza B (B/Hong Kong/CUHK50947/2004) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40709V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Hong Kong/CUHK50947/2004) hemagglutinin (AFP83206) (Met1-Tyr552), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3630