Learn More
Sino Biological Influenza B (B/Ann Arbor/1/1986) Hemagglutinin / HA Protein (His Tag)
Supplier:  Sino Biological 40706V08H100
Encompass_Preferred
A DNA sequence encoding the Influenza B virus (B/Ann Arbor/1/1986) hemagglutinin (ABF21279) (Met1-Tyr553), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3633