Learn More
Sino Biological Influenza B Hemagglutinin / HA Antibody, Mouse MAb
Supplier:  Sino Biological 11053MM09100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant influenza B virus HA (rvHA; Catalog#11053-V08H; ACA33493.1; Met 1-Ala 555). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-228-2064