Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Influenza B (Malaysia/2506/04) Culture Fluid
SDP

Catalog No. 22158007 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-007 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-007 Supplier ZeptoMetrix Supplier No. 0810258CF
Add to Cart
Add to Cart

For research, product development testing, or quality assurance testing

  • Propagated in the MDCK cell line
  • Each lot is assayed for its Tissue Culture Infective Dose (TCID50) and sold with titers >105U/mL

Specifications

Biosafety Level 2
Certifications/Compliance ISO 13485
Content And Storage -65°C or below
Form Solution
Product Type Influenza B Virus Infectious Culture Fluid
For Use With (Application) Nucleic Acid/Molecular Testing, LOD studies, Cross-reactivity studies
Packaging Type Vial
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.