Learn More
Sino Biological Influenza B NP ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370905
Encompass_Preferred
Each vial contains 210 ng of recombinant Influenza B (B/Florida/4/2006) Nucleoprotein / NP. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 8000 pg/mL is recommended.
Catalog No. 50-237-0905 Supplier Sino Biological Supplier No. SEK00715
Each vial contains 210 ng of recombinant Influenza B (B/Florida/4/2006) Nucleoprotein / NP. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 8000 pg/mL is recommended.