Learn More
Sino Biological Influenza B NP ELISA Pair Set 15PP (Sino Biological)
Supplier:  Sino Biological SEK00515
Encompass_Preferred
Each vial contains 85 ng of recombinant Influenza B (B/Washington/02/2019) Nucleoprotein. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 2500 pg/mL is recommended.
Catalog No. 50-237-1712